missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTLA Monoclonal Antibody (OTI3A12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812577S
This item is not returnable.
View return policy
Description
BTLA Monoclonal antibody specifically detects BTLA in Human samples. It is validated for Flow CytometrySpecifications
| BTLA | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q7Z6A9 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 151888 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry | |
| OTI3A12 | |
| Unconjugated | |
| BTLA | |
| A630002H24; B and T lymphocyte associated; B and T lymphocyte associated protein; B and T lymphocyte attenuator; B- and T-lymphocyte attenuator; B- and T-lymphocyte-associated protein; BTLA; BTLA_HUMAN; BTLA1; CD272; CD272 antigen; FLJ16065; MGC129743 | |
| Human recombinant protein fragment corresponding to amino acids 31-157+Mouse Fc of human BTLA produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction