missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTN1A1/Butyrophilin Antibody (OTI7B5), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70299H
This item is not returnable.
View return policy
Description
BTN1A1/Butyrophilin Monoclonal specifically detects BTN1A1/Butyrophilin in Human, Canine samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| BTN1A1/Butyrophilin | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| BTN1A1 | |
| Full length recombinant protein of human BTN1A1/Butyrophilin (NP_001723) produced in HEK293T cells. | |
| 0.1 mL | |
| 696.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI7B5 | |
| Western Blot, Immunohistochemistry | |
| BTBTNbK14H9.2 (butyrophilin, subfamily 1, member A1), butyrophilin subfamily 1 member A1, butyrophilin, subfamily 1, member A1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Canine | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction