missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTN1A1/Butyrophilin Antibody (OTI7B5), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70299MFV500
This item is not returnable.
View return policy
Description
BTN1A1/Butyrophilin Monoclonal antibody specifically detects BTN1A1/Butyrophilin in Human, Canine samples. It is validated for Flow Cytometry
Specifications
| BTN1A1/Butyrophilin | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 696 | |
| Store at 4C in the dark. | |
| IgG2a |
| Flow Cytometry | |
| OTI7B5 | |
| Flow Cytometry | |
| BTBTNbK14H9.2 (butyrophilin, subfamily 1, member A1), butyrophilin subfamily 1 member A1, butyrophilin, subfamily 1, member A1 | |
| Full length human recombinant protein of human BTN1A1 (NP_001723) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human, Canine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction