Learn More
BTN1A1 Monoclonal Antibody (OTI1A6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA501531
Encompass_Preferred
 View more versions of this product
Product Code. 17524501
  • POA / 100µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
BTN1A1 (Butyrophilin) is the major protein associated with fat droplets in the milk. It is a member of the immunoglobulin superfamily. It may have a cell surface receptor function. The human butyrophilin gene is localized in the major histocompatibility complex (MHC) class I region of 6p and may have arisen relatively recently in evolution by the shuffling of exons between 2 ancestral gene families.
Specifications
Specifications
BTN1A1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q13410
BTN1A1
Full length human recombit protein of human BTN1A1 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Immunocytochemistry, Immunofluorescence, Western Blot
OTI1A6
Unconjugated
BTN1A1
BT, BTN, BTN1
Mouse
Affinity Chromatography
RUO
696
-20° C, Avoid Freeze/Thaw Cycles