Learn More
BTN1A1 Monoclonal Antibody (OTI1A6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501531
This item is not returnable.
View return policy
Description
BTN1A1 (Butyrophilin) is the major protein associated with fat droplets in the milk. It is a member of the immunoglobulin superfamily. It may have a cell surface receptor function. The human butyrophilin gene is localized in the major histocompatibility complex (MHC) class I region of 6p and may have arisen relatively recently in evolution by the shuffling of exons between 2 ancestral gene families.Specifications
| BTN1A1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q13410 | |
| BTN1A1 | |
| Full length human recombit protein of human BTN1A1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI1A6 | |
| Unconjugated | |
| BTN1A1 | |
| BT, BTN, BTN1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 696 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |