Learn More
BTN1A1 Mouse anti-Canine, Human, Clone: OTI7B5, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF501529
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
BTN1A1 (Butyrophilin) is the major protein associated with fat droplets in the milk. It is a member of the immunoglobulin superfamily. It may have a cell surface receptor function. The human butyrophilin gene is localized in the major histocompatibility complex (MHC) class I region of 6p and may have arisen relatively recently in evolution by the shuffling of exons between 2 ancestral gene families.Specifications
| BTN1A1 | |
| Monoclonal | |
| Unconjugated | |
| BTN1A1 | |
| BT, BTN, BTN1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 100855656, 696 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI7B5 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q13410 | |
| BTN1A1 | |
| Full length human recombit protein of human BTN1A1 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Canine, Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.