missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTN2A1 Antibody (ICT-01) [Janelia Fluor™ 669], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28456JF669
This item is not returnable.
View return policy
Description
BTN2A1 Monoclonal antibody specifically detects BTN2A1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| BTN2A1 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| BK14H9.1, BTF1BT2.1butyrophilin BTF1, butyrophilin subfamily 2 member A1, butyrophilin, subfamily 2, member A1, DJ3E1.1, FLJ36567 | |
| BTN1A1 | |
| 0.1 mL | |
| Endocrinology, Signal Transduction | |
| 11120 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| ICT-01 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction