missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ BTN2A1 Antibody (ICT-01-N297A) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
€ 213.00 - € 326.00
Specifications
| Antigen | BTN2A1 |
|---|---|
| Clone | ICT-01-N297A |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30044881
|
Novus Biologicals™
NBP3-28457-100ug |
100 μg |
€ 326.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30048808
|
Novus Biologicals™
NBP3-28457-50ug |
50 μg | |||||||
|
30045999
|
Novus Biologicals™
NBP3-28457-1mg |
1 mg |
N/A
|
N/A | |||||
Description
BTN2A1 Monoclonal antibody specifically detects BTN2A1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpecifications
| BTN2A1 | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Endocrinology, Signal Transduction | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| BK14H9.1, BTF1BT2.1butyrophilin BTF1, butyrophilin subfamily 2 member A1, butyrophilin, subfamily 2, member A1, DJ3E1.1, FLJ36567 | |
| BTN1A1 | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
| ICT-01-N297A | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Q13410 | |
| 11120 | |
| IgG1 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title