missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTN3A1/2/3 Antibody (849203), PE/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB7136PECY7
This item is not returnable.
View return policy
Description
BTN3A1/2/3 Monoclonal antibody specifically detects BTN3A1/2/3 in Human samples. It is validated for Flow Cytometry
Specifications
| BTN3A1/2/3 | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Human | |
| Purified |
| Flow Cytometry | |
| 849203 | |
| Flow Cytometry | |
| BT3.1;BTF5;BTN3.1;BTN3A1;CD277 | |
| Chinese hamster ovary cell line CHO-derived recombinant human CD277/BTN3A1, Met1-Gly254, Accession # O00481 | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction