missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTN3A2 Antibody (OTI1A6), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71678PE
This item is not returnable.
View return policy
Description
BTN3A2 Monoclonal antibody specifically detects BTN3A2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| BTN3A2 | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 11118 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| OTI1A6 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
| BT3.2, BT3.3, BTF3, BTF4butyrophilin protein, butyrophilin subfamily 3 member A2, butyrophilin, subfamily 3, member A2, FLJ40011 | |
| Full length human recombinant protein of human BTN3A2 (NP_008978) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction