missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTNL2/Butyrophilin-like 2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 204.00 - € 463.00
Specifications
| Antigen | BTNL2/Butyrophilin-like 2 |
|---|---|
| Dilution | Western Blot 1:1000 - 1:5000, Immunohistochemistry 1:100 - 1:500, Immunohistochemistry-Paraffin 1:100 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18659161
|
Novus Biologicals
NBP2-92488-0.02ml |
0.02 mL |
€ 204.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18630412
|
Novus Biologicals
NBP2-92488-0.1ml |
0.1 mL |
€ 463.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BTNL2/Butyrophilin-like 2 Polyclonal antibody specifically detects BTNL2/Butyrophilin-like 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| BTNL2/Butyrophilin-like 2 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Immunology | |
| PBS with 50% glycerol, pH7.3. | |
| 56244 | |
| IgG | |
| Affinity purified |
| Western Blot 1:1000 - 1:5000, Immunohistochemistry 1:100 - 1:500, Immunohistochemistry-Paraffin 1:100 - 1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| butyrophilin-like 2 (MHC class II associated) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 246-455 of human BTNL2 (NP_062548.1). LKVNGPSQPILVRVGEDIQLTCYLSPKANAQSMEVRWDRSHRYPAVHVYMDGDHVAGEQMAEYRGRTVLVSDAIDEGRLTLQILSARPSDDGQYRCLFEKDDVYQEASLDLKVVSLGSSPLITVEGQEDGEMQPMCSSDGWFPQPHVPWRDMEGKTIPSSSQALTQGSHGLFHVQTLLRVTNISAVDVTCSISIPFLGEEKIATFSLSGW | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title