missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bu-1a Monoclonal Antibody (21-1A4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA528755
This item is not returnable.
View return policy
Description
Bu-1a Monoclonal Antibody for Flow
Bu-1, a product of the Bu-1a and Bu-1b alleles, is a type I transmembrane glycosylated disulfide-linked homodimer. The monoclonal antibody AV20 recognizes a monomorphic determinant on the Bu-1 B cell-associated alloantigens of both RPL 6(3) (Bu-1a) and 7(2) (Bu-1b) lines of inbred chickens. The antibody also identifies B cells in partially inbred birds. Bu-1 is found on 85-90% of bursal cells, 2-8% of thymocytes, 15-27% of spleen cells, and 2-18% of peripheral blood cells. It is also expressed on subsets of macrophages and monocytes but not on granulocytes, erythrocytes, or thrombocytes. The monoclonal antibody AV20 does not cross react with turkey cells when analyzed by flow cytometry.
Specifications
| Bu-1a | |
| Monoclonal | |
| 0.5 mg/mL | |
| BBS with Proprietary stabilizer and no preservative; pH 8.2 | |
| 0 | |
| LOC396098 | |
| Bursa cells from one-day-old H.B15 (Bu-1a/b) strain chickens. | |
| 1 mL | |
| Primary | |
| Chicken | |
| Antibody | |
| IgG1 κ |
| Flow Cytometry | |
| 21-1A4 | |
| Unconjugated | |
| LOC396098 | |
| B6.1; B6.2; Bu-1; Bu-1b; ChB6.1; LOC396098 | |
| Mouse | |
| Purified | |
| RUO | |
| 396098 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction