missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BUB1B Mouse anti-Human, Clone: OTI11G2, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA500540
This item is not returnable.
View return policy
Description
BUBR1 is a member of the Ser/Thr protein kinase family. It is a component of the mitotic checkpoint that delays anaphase until all chromosome are properly attached to the mitotic spindle. Compromised spindle checkpoints are thought to play an important role in genetic instability that predisposes cells to maligt transformations. Human cells contain two related checkpoint protein kinases, evolved from the BUB1 gene; hBUBR1 and hBUB1. They are important components of the spindle checkpoint Both kinases monitor mitotic kinetochore-microtubule interactions. hBUBR1 is essential for normal mitotic progression as it prevents cells from entering into anaphase too early. BUBR1 has been detected in human cancers. It has been identified as a novel binding partner of BCSG1 (Breast Cancer-Specific Gene 1), which has been suggested to accelerate the progression of breast cancer by compromising the mitotic checkpoint control through inactivation of BUBR1.Specifications
| BUB1B | |
| Monoclonal | |
| Unconjugated | |
| BUB1B | |
| BUB1beta, BUBR1, Bub1A, MAD3L, MVA1, SSK1, hBUBR1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 701 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI11G2 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O60566 | |
| BUB1B | |
| Full-length protein expressed in 293T cell transfected with human BUB1B expression vector | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction