missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bub3 Antibody (2H6), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42680B
This item is not returnable.
View return policy
Description
Bub3 Monoclonal specifically detects Bub3 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| Bub3 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| BUB3 | |
| Recombinant human BUB3 (1-328aa) purified from E. coli | |
| 100 μL | |
| Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
| 9184 | |
| Store at 4C in the dark. | |
| IgG2b κ |
| Western Blot, ELISA | |
| 2H6 | |
| Western Blot, ELISA | |
| BUB3 (budding uninhibited by benzimidazoles 3, yeast) homolog, BUB3 budding uninhibited by benzimidazoles 3 homolog, BUB3L, budding uninhibited by benomyl, budding uninhibited by benzimidazoles 3 homolog (yeast), hBUB3, mitotic checkpoint component, mitotic checkpoint protein BUB3 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion