missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bub3 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 588.00
Specifications
| Antigen | Bub3 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 450 SE |
| Form | Purified |
Description
Bub3 Polyclonal antibody specifically detects Bub3 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
| Bub3 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| BUB3 (budding uninhibited by benzimidazoles 3, yeast) homolog, BUB3 budding uninhibited by benzimidazoles 3 homolog, BUB3L, budding uninhibited by benomyl, budding uninhibited by benzimidazoles 3 homolog (yeast), hBUB3, mitotic checkpoint component, mitotic checkpoint protein BUB3 | |
| Antibody was raised against a 16 amino acid synthetic peptide from near the carboxy terminus of human bub3. The immunogen is located within the last 50 amino acids of Bub3. Amino Acid Squence: SYMYEMDDTEHPEDG | |
| Primary | |
| Store at 4°C in the dark. |
| Western Blot | |
| mFluor Violet 450 SE | |
| Rabbit | |
| Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
| 50mM Sodium Borate | |
| 9184 | |
| IgG | |
| Peptide affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title