Learn More
Invitrogen™ Bub3 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA578889
Product Code. 15954465
  • € 470.00 / 100µg
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: HELA whole cell, A549 whole cell, JURKAT whole cell, COLO320 whole cell. IHC: Rat Testis Tissue, Human Mammary Cancer Tissue IHC-F: rat lung tissue. ICC/IF: A549 Cell, HELA Cell.
This gene encodes a protein involved in spindle checkpoint function. The encoded protein contains four WD repeat domains and has sequence similarity with the yeast BUB3 protein. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Bub3
Polyclonal
Unconjugated
BUB3
Aa2-050; AU019800; AU021329; AU043350; AW146323; bub3; BUB3 budding uninhibited by benzimidazoles 3 homolog; BUB3 mitotic checkpoint protein; BUB3, mitotic checkpoint protein; BUB3L; Bub3-PA; Bub3-PB; budding uninhibited by benomyl; budding uninhibited by benzimidazoles 3 homolog; budding uninhibited by benzimidazoles 3 homolog (S. cerevisiae); C78067; CG7581; CG7581-PA; CG7581-PB; dBUB3; Dmbub3; Dmel\CG7581; Dmel_CG7581; hBUB3; mitotic checkpoint component; Mitotic checkpoint protein BUB3; OTTHUMP00000020682; testicular tissue protein Li 27; WD repeat type I transmembrane protein A72.5
Rabbit
Antigen affinity chromatography
RUO
12237, 361662, 9184
-20°C
Lyophilized
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
500 μg/mL
PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
O43684, Q9WVA3
BUB3
A synthetic peptide corresponding to a sequence in the middle region of human Bub3 (76-90aa DHQLKMHDLNTDQEN).
100 μg
Primary
Human, Mouse, Rat