Learn More
Manutan Bubble anti-static Aircap, roll width: 500 mm, roll length: 150 m
Brand:  Manutan A027100
Encompass_Preferred
Bubble anti-static Aircap, roll width: 500 mm, roll length: 150 m
Product Code. 15639291
  • € 115.56 / Pack of 2
Only null left
Estimated Shipment: 20-02-2026