missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Capsules pH 10, Reagecon™
Shop All Reagecon Products

Click to view available options
Quantity:
50 Capsules
Unit Size:
Pack of 50
Specifications
Specifications
| Color | Blue |
| pH | 10 |
| Quantity | 50 Capsules |
Safety and Handling
Product Identifier
- pH BUFFER CAPSULES pH 10.00 ± 0.02 a 25°C
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : sodium carbonate monohydrate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction