missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Elution Buffer ELU

Product Code. 11942192
Click to view available options
Quantity:
600 mL
Unit Size:
600mL
This item is not returnable. View return policy

Product Code. 11942192

Brand: Macherey-Nagel™ 740316.600

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Elution Buffer ELU to be used with NucleoBond Xtra Midi / Maxi kits.

Specifications

Quantity 600 mL
compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contains : 2-propanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.