missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution pH 10, Violet, Chem Lab™

Catalog Number 10571373
Click to view available options
Quantity:
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy
Melting Point 0°C
Color Violet
pH 10
Physical Form Liquid
Quantity 1 L
Solution Type Buffer
Formulation H3BO3-KCl-NaOH
Odor Odorless
Solubility Information Water Soluble
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'