missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 13.000, NIST, Chem-Lab

Catalog Number 11935916
Click to view available options
Quantity:
10 L
Packaging:
PE
This item is not returnable. View return policy
This item is not returnable. View return policy

Glycin-NaCl-NaOH buffer solution for laboratory use and pH measurements, traceable to SRM from NIST

Density 1.01g/mL
pH 13.00
Quantity 10 L
Formulation Glycin, NaCl, NaOH
Packaging PE
Traceability to NIST NIST traceable
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'