missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution, pH 2.0 (20°C), Honeywell™ Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 mL
6 x 500 mL
Unit Size:
500mL
Pack of 6
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| pH | 2.0 |
| Quantity | 500 mL |
| UN Number | UN1789 |
| IUPAC Name | azanium;hydroxide |
| Chemical Name or Material | Buffer solution |
Safety and Handling
Product Identifier
- Buffer solution pH 2.0
- Warning
- Substances and mixtures corrosive to metals Category 1
- H290-May be corrosive to metals.
- P234-Keep only in original container.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- MIXTURE LIST-Contains : hydrochloric acid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction