Learn More
Buffer Solution, pH 4.64, METTLER TOLEDO™
Shop All Mettler Toledo Products
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
Specifications
Specifications
| Color | Colorless |
| pH | 4.64 |
| Physical Form | Liquid |
| Quantity | 250 mL |
| Solution Type | pH Buffer |
| Odor | None |
| Packaging | 250mL |