Learn More
Color Coded pH Buffers in Twin Neck Bottle Format, Reagecon™
Shop All Reagecon Products
Click to view available options
Quantity:
250 mL
500 mL
1 L
pH:
10 at 20°C
10 at 25°C
7 at 20°C
7 at 25°C
Förpackningsstorlek
1 L
250mL
500mL
Beskrivelse
- Colored pH Buffer Solutions
- Tested at 20°C or 25°C
- Certified by Reagecon’s ISO/IEC 17025 AccreditedTest Method
- NIST traceable
- Presented in various pack sizes
- No possibility of contamination
- No need for separate measuring container for use in the calibration of the Electrode
- Correct quantity of buffer required for calibration is dispensed into the calibrating chamber giving rise to no waste
- Ideally suited for field work
- Easy to carry
- 250ml, 500ml and 1L sizes available
Tekniske data
Tekniske data
| Color | Yellow |
| pH | 7 at 20°C |
| Quantity | 1 L |
| Packaging | HDPE Twin Neck Bottle |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion