Learn More
Color Coded pH Buffers in Twin Neck Bottle Format, Reagecon™
BIOPHARMA
Product Code. 12037365 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
pH:
10 at 20°C
10 at 25°C
7 at 20°C
7 at 25°C
Unit Size:
1 L
250mL
500mL
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity pH unitSize
12037365 250 mL 7 at 20°C 250mL
11434599 250 mL 10 at 25°C 250mL
11454589 250 mL 10 at 20°C 250mL
11404599 250 mL 7 at 25°C 250mL
11444599 500 mL 10 at 25°C 500mL
11802172 500 mL 7 at 25°C 500mL
11464589 1 L 10 at 20°C 1 L
11454599 1 L 10 at 25°C 1 L
11444589 1 L 7 at 20°C 1 L
11424599 1 L 7 at 25°C 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 12037365 Supplier Reagecon Supplier No. 10702CTT
Use color coded pH buffers for ease of use. Reagecon™ Color Coded pH Buffer in a Twin Neck Bottle Format are tested at 20°C or 25°C and are certified and NIST traceable.
  • Colored pH Buffer Solutions
  • Tested at 20°C or 25°C 
  • Certified by Reagecon’s ISO/IEC 17025 AccreditedTest Method
  • NIST traceable
  • Presented in various pack sizes
Twin Neck Bottle Advantages
  • No possibility of contamination
  • No need for separate measuring container for use in the calibration of the Electrode
  • Correct quantity of buffer required for calibration is dispensed into the calibrating chamber giving rise to no waste
  • Ideally suited for field work
  • Easy to carry
  • 250ml, 500ml and 1L sizes available
Specifications
Color Yellow
pH 7 at 20°C
Quantity 250 mL
Packaging HDPE Twin Neck Bottle