Learn More
Vitlab™ Bulb Pipettes
Vitlab™ Bulb Pipettes offer higher accuracy when measuring liquid volumes.
€ 157.98 - € 166.56
Specifications
Class B
Description Fixed Volumetric Pipets
Material Polypropylene
Temperature (Metric) Operating <60°C
View More Specs
Products 2
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
10224972
View Documents Promotion Details
Vitlab™
164294
5 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10530273
View Documents Promotion Details
Vitlab™
164394
10 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
  • Calibrated to deliver ‘Ex’
  • Highly transparent
  • With high-contrast, blue markings
  • Break-resistant
  • High chemical resistance
  • Exposure to temperatures above 60°C can lead to volume changes
  • Recommended cleaning with mild alkaline detergents up to 60°C
Specifications
Specifications
B
Fixed Volumetric Pipets
Polypropylene
<60°C