Learn More
Deutsch Neumann™ The Flip™ Safety Pipette Fillers
Reduces the risks of pipetting when handling toxic, corrosive, bacteria contaminated or bad smelling liquids. Deutsch Neumann™ The Flip™ Safety Pipette Fillers are manufactured from natural rubber.
Brand: Deutsch Neumann™ 3544450
Additional Details : Weight : 0.03600kg
Description
- Only two operating points
- Evacuate via the new automatic valve
- One-handed design is the simplest safety pipette available
- Ability to clean inside of bulb by removing patented valve and rinsing out