Learn More
Deutsch Neumann™ The Flip™ Safety Pipette Fillers
Reduces the risks of pipetting when handling toxic, corrosive, bacteria contaminated or bad smelling liquids. Deutsch Neumann™ The Flip™ Safety Pipette Fillers are manufactured from natural rubber.
Brand:  Deutsch Neumann™ 3544450
Additional Details : Weight : 0.03600kg
Product Code. 10441021
  • € 28.10 / Each
In Stock
to see stock.
Description
Description
  • Only two operating points
  • Evacuate via the new automatic valve
  • One-handed design is the simplest safety pipette available
  • Ability to clean inside of bulb by removing patented valve and rinsing out