Learn More
Trajan Scientific and Medical™ Bulkhead Unions
Gives low dead volume connections for optimum chromatography.
Brand:  Trajan Scientific and Medical™ 101340
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15229103
  • € 59.60 / Each
Estimated Shipment: 07-06-2024
to see stock.
Add to basket
Description
Description
  • Low dead volume
  • High temperature applications
  • Leak free connections
Specifications
Specifications
Bulkhead Union
Size: 1/16 x 1/16 in.