Learn More
Aalborg™ Buna N O-ring Aluminum Frame with Plug
Product Code. 11768528
Change view
Click to view available options
Size:
2-Tube, 150 mm
3-Tube, 150 mm
4-Tube, 150 mm
5-Tube, 150 mm
Includes:
Plug
Plug and Manifold
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Includes unitSize
11768528 2-Tube, 150 mm Plug Each
11778528 2-Tube, 150 mm Plug and Manifold Each
11748548 4-Tube, 150 mm Plug Each
11753189 4-Tube, 150 mm Plug and Manifold Each
11753349 5-Tube, 150 mm Plug and Manifold Each
11728538 3-Tube, 150 mm Plug Each
11738538 3-Tube, 150 mm Plug and Manifold Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 11768528 Supplier Aalborg™ Supplier No. P21A2BA0
  • ONLINE EXCLUSIVE PRICE
    € 288.75
    € 175.44 / Pack of 1
    Save € 113.31
    39% Off
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
Aalborg™ Buna N O-ring Aluminum Frame with Plug is for use with multiple tube flow meters, which retain most of the unique features associated with single tube units.
  • Rib-guided or fluted metering tubes facilitate stable, accurate readings
  • Interchangeability of flow tubes and floats
  • Manifolding at inlet or outlet
  • Ease of installation and exchange of flow tubes
  • Chemical compatibility
  • Simple means of mounting
Specifications
For Use With (Equipment) Gas/Liquid Flowmeter (Parallel-mount)
Type Frame
Includes Plug
Size 2-Tube, 150 mm