Learn More
Kartell™ Burette Clamp
Click to view available options
:
Includes: 2 Clamps
Description
Kartell Burette Clamp, Includes: 2 Clamps, Polypropylene, Buret Clamp, White, No. per Pack: 1 Each
Specifications
Specifications
| Color | White |
| Material | Polypropylene |
| No. per Pack | 1 Each |
| Includes | (2) Clamps |
| Type | Buret Clamp |
| Diameter (Metric) Rod | 8 to 14 mm |