Learn More
Buerkle™ Mercury Collector
Product Code. 10483482 Shop All Bürkle Products
Click to view available options
:
Includes: Special closure with special plastic foam
Product Code. 10483482
Brand: Buerkle™ 97890001
Only null left
Add to basket
Add to basket
Buerkle™ Mercury Collector enables the easy collection of spilt mercury, which can then be used again.
  • Enables the easy collection of spilt mercury, which can then be used again
  • The mercury collector is composed of a tub with a screw cap lid that has a special plastic foam inserted
  • On pressing this foam gently on the spilt mercury, the foam's pores are opened and the mercury globules are absorbed
  • The cap is then screwed on the tub, the globules are released again and fall through the perforated separation into the tub
TRUSTED_SUSTAINABILITY
Specifications
Type Mercury Collector
For Use With (Application) Mercury
Includes Special closure with special plastic foam