Learn More
Buspirone.HCl, 1mg/ml in Methanol (as free base), Lipomed
Product Code. 30461788
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30461788 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30461788 Supplier Lipomed Supplier No. LPMBSP1832HC1LM
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.
Buspirone.HCl, 1mg/ml in Methanol (as free base)
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Buspirone hydrochloride
CAS 33386-08-2
Analyte or Component Names Buspirone hydrochloride
Shipping Condition Room Temperature
Molecular Formula C21 H31 N5 O2 . Cl H
Concentration 1 mg/mL
Synonym Buspirone Hydrochloride
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES Cl.O=C1CC2(CCCC2)CC(=O)N1CCCCN3CCN(CC3)c4ncccn4
IUPAC Name 8-[4-(4-pyrimidin-2-ylpiperazin-1-yl)butyl]-8-azaspiro[4.5 ]decane-7,9-dione;hydrochloride
Solvent or Matrix Methanol
Industry Type Pharmaceutical
Molecular Weight (g/mol) 421.96
Quantity 1 mL
Formula Weight 421.2245 g/mol