Learn More
Buspirone Hydrochloride, TRC
Product Code. 30134243
Change view
Click to view available options
Quantity:
50 mg
1 g
5 g
Unit Size:
1g
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134243 5 g 5g
18061350 1 g 1g
18002550 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30134243 Supplier Toronto Research Chemicals Supplier No. TRCB6898505G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 33386-08-2
Molecular Formula C21 H31 N5 O2 . Cl H
Molecular Weight (g/mol) 421.96
Synonym Buspirone Hydrochloride, 8-Azaspiro[4.5]decane-7,9-dione, 8-[4-[4-(2-pyrimidinyl)-1-piperazinyl]butyl]-, hydrochloride (1:1) (ACI), 1,1-Cyclopentanediacetimide, N-[4-[4-(2-pyrimidinyl)-1-piperazinyl]butyl]-, monohydrochloride (8CI), 8-Azaspiro[4.5]decane-7,9-dione, 8-[4-[4-(2-pyrimidinyl)-1-piperazinyl]butyl]-, monohydrochloride (9CI), 8-[4-[4-(2-Pyrimidinyl)-1-piperazinyl]butyl]-8-azaspiro[4.5]decane-7,9-dione hydrochloride, Ansial, Ansiced, Ansiten, Axoren, Bespar, Buscalma, Buspa, Buspar, Buspimem, Buspinol, Buspirone hydrochloride, Buspirone monohydrochloride, Buspisal, Busron, Censpar, Lucelan, MJ 9022-1, Narol, Travin
IUPAC Name 8-[4-(4-pyrimidin-2-ylpiperazin-1-yl)butyl]-8-azaspiro[4.5]decane-7,9-dione;hydrochloride
SMILES Cl.O=C1CC2(CCCC2)CC(=O)N1CCCCN3CCN(CC3)c4ncccn4
Specifications
Quantity 5 g
InChI Formula InChI=1S/C21H31N5O2.ClH/c27-18-16-21(6-1-2-7-21)17-19(28)26(18)11-4-3-10-24-12-14-25(15-13-24)20-22-8-5-9-23-20;/h5,8-9H,1-4,6-7,10-17H2;1H
Formula Weight 421.2245
Chemical Name or Material Buspirone hydrochloride