Learn More
Butylphthalide, MedChemExpress
Product Code. 16424437
Encompass
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16424437 50 mg 50mg
16408568 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16424437 Supplier Medchem Express Supplier No. HYB0647
MedChemExpress Butylphthalide(3-n-Butylphthalide) is an anti-cerebral-ischemia drug; first isolated from the seeds of celery, showed efficacy in animal models of stroke.
Butylphthalide(3-n-Butylphthalide) is an anti-cerebral-ischemia drug
Specifications
Chemical Name or Material Butylphthalide
Physical Form Liquid
CAS 6066-49-5
Color Colorless
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C12H14O2
For Use With (Application) Neuroscience-Neuromodulation
Synonym 3-n-Butylphthalide 3-Butylphthalide
Solubility Information DMSO : 250 mg/mL (1314.13 mM; Need ultrasonic)
SMILES O=C1OC(CCCC)C2=C1C=CC=C2
Molecular Weight (g/mol) 190.24
Formula Weight 190.24
Percent Purity 98.17%
Grade Research
Show More Show Less
Research purposes only