Learn More
Butyraldehyde 2,4-Dinitrophenylhydrazone-d3, TRC
Specifications
Specifications
| Chemical Name or Material | Butyraldehyde 2,4-Dinitrophenylhydrazone-d3 |
| CAS | 259824-54-9 |
| Molecular Formula | C10 D3 H9 N4 O4 |
| Quantity | 5 mg |
| Synonym | Butanal, (4,6-dinitrophenyl-2,3,5-d3)hydrazone (9CI), Butanal 2-(2,4-dinitrophenyl-d3)hydrazone, Butyraldehyde-2,4-dinitrophenylhydrazone-d3, Butyraldehyde-2,4-dinitrophenylhydrazone-D3 (phenyl-D3) |
| InChI Formula | InChI=1S/C10H12N4O4/c1-2-3-6-11-12-9-5-4-8(13(15)16)7-10(9)14(17)18/h4-7,12H,2-3H2,1H3/i4D,5D,7D |
| SMILES | [2H]c1c([2H])c(c([2H])c(c1NN=CCCC)[N+](=O)[O-])[N+](=O)[O-] |
| IUPAC Name | N-(butylideneamino)-2,3,5-trideuterio-4,6-dinitroaniline |
| Molecular Weight (g/mol) | 255.245 |
| Formula Weight | 255.105 |