Learn More
Butyric-2,2-d2 Acid, TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | Butyric-2,2-d2 Acid |
| CAS | 19136-92-6 |
| Molecular Formula | C4 D2 H6 O2 |
| Quantity | 500 mg |
| Synonym | Butanoic-2,2-d2 acid (9CI), Butyric-2,2-d2 acid (8CI), Butanoic acid 2,2-D2 |
| InChI Formula | InChI=1S/C4H8O2/c1-2-3-4(5)6/h2-3H2,1H3,(H,5,6)/i3D2 |
| SMILES | [2H]C([2H])(CC)C(=O)O |
| IUPAC Name | 2,2-dideuteriobutanoic acid |
| Molecular Weight (g/mol) | 90.117 |
| Formula Weight | 90.065 |