missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butyric acid hydrazide, 95%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
Specifications
Melting Point | 43.0°C to 44.0°C |
Boiling Point | 120.0°C (10.0 mmHg) |
Infrared Spectrum | Authentic |
Assay Percent Range | 94% min |
Linear Formula | CH3CH2CH2C(O)NHNH2 |
Quantity | 5 g |
Solubility Information | Solubility in water: soluble. |
Formula Weight | 102.14 |
Percent Purity | 95% |
Chemical Name or Material | Butyric acid hydrazide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'