missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BVES Polyclonal Antibody
GREENER_CHOICE

Product Code. 17137393
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17137393 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17137393 Supplier Invitrogen™ Supplier No. PA5109460

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: SEPFLYEIFR YLIGKDITNK LYSLNDPTLN DKKAKKLEHQ LSLCTQISML EMRNSIASSS DSDDGLHQFL RGTSSMSSLH VSSPHQRASA KMKPIEEGAE DDDDVFEPAS PNTLKVHQLP.

Blood vessel epicardial substance (BVES), also designated Popeye protein 1, is a transmembrane protein that plays a role in cell-cell interactions and adhesion, specifically at tight junctions. BVES is composed of an extracellular amino-terminus, three transmembrane domains and a cytoplasmic carboxyl-terminus. It is expressed in the developing coronary vascular system, specifically in the proepicardium, migrating epithelial epicardium, delaminated vasculogenic mesenchyme and vascular smooth muscle cells, where it functions to direct development in heart, epithelial and muscle cells during embryogenesis. BVES accumulates at points of cell-cell contact, such as filopodia and cell borders, and promotes adhesion prior to the arrival of E-cadherin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BVES
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene BVES
Gene Accession No. Q8NE79
Gene Alias blood vessel epicardial substance; bves; hBVES; LGMD2X; mBVES; MGC42413; POP 1; POP1; POPDC 1; Popdc1; popdc1 protein; popeye 1; popeye domain containing 1; popeye domain-containing 1; popeye domain-containing protein 1; Popeye protein 1; zgc:86887
Gene Symbols BVES
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 241-360 of human BVES (NP_001186492.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11149
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.