missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ BVT 948

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
BVT 948 is a non-competitive, cell-permeable inhibitor of protein tyrosine phosphatases (PTPs) (IC50 = 0.09 - 1.7 μM); displays irreversible inhibition through catalysis of the hydrogen peroxide-dependent oxidation of PTP. Enhances insulin signaling in vitro and insulin tolerance in ob/ob mice in vivo. Also inhibits several cytochrome P450 isoforms (IC50 <10 μM).
Specifications
Specifications
| Quantity | 50 mg |
| Product Type | BVT 948 |
| Purity | >98% |
| Molecular Weight (g/mol) | 241.25 |
| Inhibitors | BVT 948 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction