missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ BX 430
GREENER_CHOICE

Product Code. 18759708
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18759708

Brand: Tocris Bioscience™ 5545/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective P2X4 allosteric antagonist

BX 430 is a selective P2X4 allosteric antagonist (IC50 = 0.54 μM). Exhibits >10-fold selectivity for human P2X4 receptors over P2X1, P2X2, P2X3, P2X5 and P2X7 receptors. Binds noncompetitively at extracellular allosteric site. Effective at human and zebrafish, but not mouse and rat P2X4 receptors.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[2,6-Dibromo-4-(1-methylethyl)phenyl]-N′-(3-pyridinyl)urea
CAS 688309-70-8
Quantity 50 mg
Target P2X Receptor Antagonists
Molecular Formula C15H15Br2N3O
Formula Weight 413.11
Purity >99%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.