missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ BX 430

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
BX 430 is a selective P2X4 allosteric antagonist (IC50 = 0.54 μM). Exhibits >10-fold selectivity for human P2X4 receptors over P2X1, P2X2, P2X3, P2X5 and P2X7 receptors. Binds noncompetitively at extracellular allosteric site. Effective at human and zebrafish, but not mouse and rat P2X4 receptors.
Specifications
Specifications
| Chemical Name or Material | N-[2,6-Dibromo-4-(1-methylethyl)phenyl]-N′-(3-pyridinyl)urea |
| CAS | 688309-70-8 |
| Quantity | 50 mg |
| Target | P2X Receptor Antagonists |
| Molecular Formula | C15H15Br2N3O |
| Formula Weight | 413.11 |
| Purity | >99% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction