Learn More
Invitrogen™ BXDC1 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA570486
Product Code. 15868893
  • € 530.00 / 100µL
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 79%; Zebrafish: 93%.
RPF2 is a protein coding gene involved in ribosomal large subunit assembly. May regulate the localization of the 5S RNP/5S ribonucleoprotein particle to the nucleolus.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
BXDC1
Polyclonal
Unconjugated
Rpf2
2810470K21Rik; AU040229; AU043242; bA397G5.4; brix domain containing 1; brix domain-containing protein 1; Brix domain-containing protein 1-like protein; BXDC1; homolog of Rpf2; hypothetical protein LOC406587; ik:tdsubc_1a3; MNCb-4285; ribosomal processing factor 2 homolog; ribosome biogenesis protein RPF2 homolog; Ribosome production factor 2 homolog; ribosome production factor 2 homolog (S. cerevisiae); RP11-397G5.1; Rpf2; Unknown (protein for MGC:134144); wu:fe17d08; wu:fj16d06; xx:tdsubc_1a3; zgc:77150
Rabbit
Affinity chromatography
RUO
294436
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
0
Rpf2
synthetic peptide directed towards the N-terminal region of rat BXDC1.