missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BXDC2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13425147
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13425147 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13425147 Supplier Invitrogen™ Supplier No. PA530646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: A549, HeLa. Predicted reactivity: Mouse (98%), Rat (98%), Xenopus laevis (88%), Pig (100%), Chicken (91%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Required for biogenesis of the 60S ribosomal subunit.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BXDC2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene Brix1
Gene Accession No. Q8TDN6
Gene Alias 1110064N10Rik; BRIX; brix domain containing 2; brix domain-containing protein 2; Brix1; BRX1, biogenesis of ribosomes; BRX1, biogenesis of ribosomes, homolog; BRX1, biogenesis of ribosomes, homolog (S. cerevisiae); BXDC2; C76935; RGD1308508; ribosome biogenesis protein Brix; ribosome biogenesis protein BRX1 homolog
Gene Symbols Brix1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human BXDC2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55299
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.