missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C/EBP gamma Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142349
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:2,000.
The C/EBP family of transcription factors regulates viral and cellular CCAAT/enhancer element-mediated transcription. C/EBP proteins contain the bZIP region, which is characterized by two motifs in the C-terminal half of the protein: a basic region involved in DNA binding and a leucine zipper motif involved in dimerization. The C/EBP family consist of several related proteins, C/EBP alpha, C/EBP beta, C/EBP gamma, and C/EBP delta, that form homodimers and that form heterodimers with each other. CCAAT/enhancer binding protein gamma may cooperate with Fos to bind PRE-I enhancer elements.
Specifications
| C/EBP gamma | |
| Polyclonal | |
| Unconjugated | |
| CEBPG | |
| C EBP; C/EBP; c/EBP gamma; C/EBP[g]; C/EBPgamma; CCAAT/enhancer binding protein (C/EBP), gamma; CCAAT/enhancer binding protein ,gamma; CCAAT/enhancer binding protein gamma; CCAAT/enhancer-binding protein gamma; CEBP gamma; CEBPG; CEBPRNA; G-CSF promoter element 1 binding protein; GPE1-binding protein; Gpe1bp; GPE1-BP; GPE1-BP (C/EBP gamma); Granulocyte colony-stimulating factor promoter element 1-binding protein; homolog to a human CCAAT/enhancer binding protein - gamma; Ig/EBP; IG/EBP-1; immunoglobulin enhancer-binding protein 1 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 1054 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P53567 | |
| CEBPG | |
| Peptide with sequence C-ENTTADGDNAGQ (aa 139-150). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction