Learn More
Ekastu Safety™ Eye Wash Bottle MINI with Funnel
Made for immediate rinsing of the eyes after accidental splashing from acids, bases and other hazardous chemicals. EKASTU Safety™ Eye Wash Bottle MINI with Funnel can be used in both upright and horizontal positions and can be carried in breast and coat bags.
Brand: Ekastu Safety™ 177259
Additional Details : Weight : 0.10000kg
Description
- Eye Wash Bottle with funnel
- DIN EN 15154-4
- Supplied empty or filled
- For immediate rinsing of the eyes after corrosion by acids, bases and other harsh chemicals
- PE bottle with screwed-on eye funnel and tilting valve
- Can be used in both upright and horizontal positions
- leak-proof
- Stores easily in EPS Eye Wash Station (sold separately)
Specifications
200 mL | |
Saline Solution | |
Eye Wash Bottle |
EN 15154-4 DIN | |
Polyethylene |