missing translation for 'onlineSavingsMsg'
Learn More

c-Fos Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

€ 186.00 - € 499.00

Specifications

Antigen c-Fos
Concentration 1 mg/mL
Dilution Western Blot 1:1000 - 1:2000, Immunohistochemistry 1:20000-1:50000, Immunocytochemistry/ Immunofluorescence 1:5000-1:20000, Immunohistochemistry Free-Floating
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Free Floating)
Classification Polyclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18640808
View Documents
Novus Biologicals
NBP2-50057
0.1 mL
€ 499.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18627908
View Documents
Novus Biologicals
NBP2-50057SS
0.025 mL
€ 186.00
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

c-Fos Polyclonal antibody specifically detects c-Fos in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry Free-Floating
Specifications

Specifications

c-Fos
Western Blot 1:1000 - 1:2000, Immunohistochemistry 1:20000-1:50000, Immunocytochemistry/ Immunofluorescence 1:5000-1:20000, Immunohistochemistry Free-Floating
Polyclonal
Purified
RUO
Human, Mouse, Rat
P01100
2353
IgG
Immunogen affinity purified
1 mg/mL
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Free Floating)
Unconjugated
Rabbit
Apoptosis, Cancer, Cellular Markers, Hypoxia, Neuronal Cell Markers, Neuroscience, Transcription Factors and Regulators, Tumor Suppressors
50% PBS, 50% glycerol
activator protein 1, AP-1, cellular oncogene c-fos, Cellular oncogene fos, C-FOS, FBJ murine osteosarcoma viral (v-fos) oncogene homolog (oncogene FOS), FBJ murine osteosarcoma viral oncogene homolog, FOS, Fos proto-oncogene, AP-1 trancription factor subunit, G0/G1 switch regulatory protein 7, G0S7, p55, proto-oncogene c-Fos
This c-Fos Antibody was developed against recombinant full length human c-Fos. [UniProt# P01100]
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.