missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-jun Antibody (5B1), Novus Biologicals™
Mouse Monoclonal Antibody
€ 200.00 - € 577.00
Specifications
| Antigen | c-jun |
|---|---|
| Clone | 5B1 |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18623236
|
Novus Biologicals
NBP2-37576-0.025ml |
0.025 mL |
€ 200.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18163499
|
Novus Biologicals
NBP2-37576 |
0.1 mL |
€ 577.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
c-jun Monoclonal specifically detects c-jun in Human, Mouse, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| c-jun | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human, Mouse, Primate, Monkey | |
| Activator protein 1, AP1, AP-1, c-Jun, enhancer-binding protein AP1, Jun activation domain binding protein, jun oncogene, jun proto-oncogene, Proto-oncogene c-Jun, transcription factor AP-1, V-jun avian sarcoma virus 17 oncogene homologp39, v-jun sarcoma virus 17 oncogene homolog, v-jun sarcoma virus 17 oncogene homolog (avian) | |
| JUN | |
| IgG1 | |
| Unpurified |
| 5B1 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, Cell Cycle and Replication, Hypoxia, Immunology, Innate Immunity, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors, Wnt Signaling Pathway | |
| Ascites with 0.03% Sodium Azide | |
| 3725 | |
| Purified recombinant fragment of human c-Jun expressed in E. Coli. | |
| Primary | |
| 43 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title