Learn More
C-Jun Monoclonal Antibody (OTI2E11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA800595
Encompass_Preferred
 View more versions of this product
Product Code. 17538602
  • € 540.87 / 100µL
Description
c-Jun is a transcription factor that recognizes and binds to the enhancer heptamer motif 5′-TGA[CG]TCA-3′. It promotes activity of NR5A1 when phosphorylated by HIPK3 leading to increased steroidogenic gene expression upon cAMP signaling pathway stimulation. Involved in activated KRAS-mediated transcriptional activation of USP28 in colorectal cancer (CRC) cells.
Specifications
Specifications
C-Jun
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P05412
JUN
Full length human recombit protein of human JUN produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Western Blot
OTI2E11
Unconjugated
JUN
AP-1, AP1, c-Jun, p39
Mouse
Affinity Chromatography
RUO
3725
-20° C, Avoid Freeze/Thaw Cycles