missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C-Jun Monoclonal Antibody (OTI2G4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800557S
This item is not returnable.
View return policy
Description
C-Jun Monoclonal antibody specifically detects C-Jun in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| C-Jun | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q5T1V6 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 83479 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2G4 | |
| Unconjugated | |
| DDX59 | |
| Activator protein 1; AH119; AP1; AP-1; Avian sarcoma virus 17 (v-jun) oncogene homolog; cjun; c-Jun; c-jun transcription factor; enhancer-binding protein AP1; I79_009953; immediate early; JUN; Jun A; Jun activation domain binding protein; Jun oncogene; jun proto-oncogene; Jun proto-oncogene, AP-1 transcription factor subunit; Junc; JUND; p39; proto-oncogene c-Jun; Rjg-9; transcription factor; transcription factor AP-1; V-jun avian sarcoma virus 17 oncogene homolog; v-jun sarcoma virus 17 oncogene homolog | |
| Human recombinant protein fragment corresponding to amino acids 1-303 of human DDX59 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction