Learn More
c-Maf Antibody (SR1746), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22154-100ul
This item is not returnable.
View return policy
Description
c-Maf Monoclonal antibody specifically detects c-Maf in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| c-Maf | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| SR1746 | |
| Western Blot 1:500-1:2000 | |
| Avian musculoaponeurotic fibrosarcoma (MAF) protooncogene, c-MAF, c-maf proto-oncogene, MGC71685, Proto-oncogene c-Maf, T lymphocyte c-maf long form, transcription factor Maf, v-maf musculoaponeurotic fibrosarcoma (avian) oncogene homolog, V-maf musculoaponeurotic fibrosarcoma oncogene homolog, v-maf musculoaponeurotic fibrosarcoma oncogene homolog (avian) | |
| A synthesized peptide derived from human c-Maf (Uniprot #: O75444) | |
| 100 μg | |
| Apoptosis, Cancer, Cell Biology, Immunology | |
| 4094 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |