missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ c-Mpl Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5105256
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total TPOR.
In 1990 an oncogene, v-mpl, was identified from the murine myeloproliferative leukemia virus that was capable of immortalizing bone marrow hematopoietic cells from different lineages. In 1992 the human homologue, named, c-mpl, was cloned. Sequence data revealed that c-mpl encoded a protein that was homologous with members of the hematopoietic receptor superfamily. Presence of anti-sense oligodeoxynucleotides of c-mpl inhibited megakaryocyte colony formation. The ligand for c-mpl, thrombopoietin, was cloned in 1994. Thrombopoietin was shown to be the major regulator of megakaryocytopoiesis and platelet formation. The protein encoded by the c-mpl gene, CD110, is a 635 amino acid transmembrane domain, with two extracellular cytokine receptor domains and two intracellular cytokine receptor box motifs. TPO-R deficient mice were severely thrombocytopenic, emphasizing the important role of CD110 and thrombopoietin in megakaryocyte and platelet formation. Upon binding of thrombopoietin CD110 is dimerized and the JAK family of non-receptor tyrosine kinases, as well as the STAT family, the MAPK family, the adaptor protein Shc and the receptors themselves become tyrosine phosphorylated.
Specifications
| c-Mpl | |
| Polyclonal | |
| Unconjugated | |
| MPL | |
| CD110; c-mpl; hlb219; MPL; MPL proto-oncogene, thrombopoietin receptor; MPLV; myeloproliferative leukemia protein; myeloproliferative leukemia virus oncogene; proto-oncogene c-Mpl; THCYT2; thrombopoietin receptor; TPOR; TPO-R | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 17480, 366455, 4352 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P40238, Q08351 | |
| MPL | |
| A synthesized peptide derived from human MPL(Accession P40238), corresponding to amino acid residues M610-P635. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction